Advantech L3-S5 Standard Sieve, 3" dia, Stainless Steel Mesh, 4.00 mm Sieve Opening, No. 5
Representative image only.
Advantech Mfr # L3-S5 – Item # EW-59948-22
Steel mesh meets ASTM and ISO specifications
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialAcrylic frame, stainless steel wire
  • U.S. Standard Screen Size (No.)5
  • Tyler Standard Screen Size (mesh)5
  • Sieve Opening (mm)4
  • Inside Diameter (InchShort)3
  • Height (InchShort)0.625
  • DescriptionStandard Sieve, Stainless Steel Mesh, 4.00 mm sieve opening, No. 5
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Standard sieves with stainless steel mesh meet ASTM and ISO specifications and are supplied with a certificate of compliance. Sieves measure 3" ID and 5/8" H.