AEMC 1000-24-1-1 Flexible Current Probe; 1000 A range, 1 mV/A output
Representative image only.
AEMC Mfr # 1000-24-1-1 – Item # EW-27747-50
Optimize performance of your electrical equiptment
  • 1 year warranty
PRODUCT OPTIONS
$432.25USD / Each
sign in for your price.
$119.00USD / Each
for an additional
$119.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • AC Min (amps)0.5
  • AC Max (amps)240
  • Max Clamp Size0.78
  • Safety RatingCAT III 1000V, CAT IV 600V, EN 61010-1
  • DescriptionFlexible Current Probe; 1000 A range, 1 mV/A output
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Use with DMMs, oscilloscopes, dataloggers, handheld scopes and power meters.