AEMC 1000-24-2-1 AmpFlex flexible current probe 100/1000 A, 24", 10 mV/1 mV/A

Representative image only.
AEMC Mfr # 2112.98 – Item # EW-20002-33
  • 1 year warranty

$437.00USD / Each
sign in for your price.

$108.00 - $178.00USD / Each

for an additional
$108.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.

SPECS & DESCRIPTION

Specifications & Description

  • AC Min (amps)100
  • AC Max (amps)1000
  • Safety RatingCAT III 1000V, CAT IV 600V, EN 61010-1
  • DescriptionAmpFlex flexible current probe 100/1000 A, 24", 10 mV/1 mV/A
  • Warranty1 Year
1 Year Warranty

Key Features

  • 1 year warranty

More About this Item

These AmpFlex ® flexible AC current probes are composed of a flexible sensor and an electronic module. The flexible sensor permits measurements on conductors where standard clamp-on probes cannot be used. In particular, it can be installed in tight spaces, around cable bundles, around wide or large bus bars, or even wrapped around irregular shapes. The Shape Memory feature enables the user to pre-shape the sensor before inserting in between or around the conductor, enhances user safety, and alleviates the drooping effect associated with other flexible sensors.

FEATURES

  • Measure from 30Arms to 30,000 Arms
  • Lengths from 24" to 60" standard
  • Accuracy 1% of read

  • REVIEWS

    Q&A

    CUSTOMER SUPPORT

    Need Help with this product?

    Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

    Call Us At1-800-323-4340
    Live chat