AEMC 3000-24-2-1 AmpFlex flexible current probe, 300/3000 A, 24", 10 mV/1 mV/A
Representative image only.
AEMC Mfr # 3000-24-2-1 – Item # EW-20002-35
  • 1 year warranty
PRODUCT OPTIONS
$479.75USD / Each
sign in for your price.
$119.00USD / Each
for an additional
$119.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • AC Min (amps)300
  • AC Max (amps)3000
  • Safety RatingCAT III 1000V, CAT IV 600V, EN 61010-1
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
These AmpFlex ® flexible AC current probes are composed of a flexible sensor and an electronic module. The flexible sensor permits measurements on conductors where standard clamp-on probes cannot be used. In particular, it can be installed in tight spaces, around cable bundles, around wide or large bus bars, or even wrapped around irregular shapes. The Shape Memory feature enables the user to pre-shape the sensor before inserting in between or around the conductor, enhances user safety, and alleviates the drooping effect associated with other flexible sensors.
FEATURES
  • Measure from 30Arms to 30,000 Arms
  • Lengths from 24" to 60" standard
  • Accuracy 1% of read
  • ACCESSORIES