AEMC 603 Clamp Meter, 1000 AC, 3000 A DC

Representative image only.
AEMC Mfr # 603 – Item # EW-20043-38
Models with Bluetooth communication capability provide additional flexibility
  • Measure’s up to 1000 V AC, DC and AC+DC with resolution to 10 mV
  • Measures single and three phase power (Real, Reactive and Apparent) up to 3 MW with resolution to 1W
  • Measures voltage frequency to 20 kHz with 0.1 Hz resolution
  • Auto selects AC or DC measurement
  • Measures Harmonics up to the 25th
  • Records up to 1000 measurements
  • Bluetooth communication and DataView® software compatible (models 407, and 607)
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • DC Max (Volts)1000
  • DC Max (amps)3000
  • AC Max (amps)2000
  • Resistance Max (KΩ)100
  • ContinuityYes (<40 Ω)
  • Continuity / Diode TestYes - Diode Test
  • Count1000
  • Current Frequency1 kHz
  • Jaw Size2.36
  • Safety RatingCAT IV 1000 V
  • PowerFour AA batteries
  • Width (in)4.375
  • Height (in)11.625
  • Depth (in)1.625
  • DescriptionClamp Meter, 1000 AC, 3000 A DC
  • Warranty1 Year
1 Year WarrantyCanadian Standards AssociationIP54 RatedUL Listed.

Key Features

  • Measure’s up to 1000 V AC, DC and AC+DC with resolution to 10 mV
  • Measures single and three phase power (Real, Reactive and Apparent) up to 3 MW with resolution to 1W
  • Measures voltage frequency to 20 kHz with 0.1 Hz resolution
  • Auto selects AC or DC measurement
  • Measures Harmonics up to the 25th
  • Records up to 1000 measurements
  • Bluetooth communication and DataView® software compatible (models 407, and 607)
  • 1 year warranty

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat