AEMC 8220 W/MN193-BK Power Quality Meter with 6A/120Aac Probe Model 8220 with MN93-Bk
Representative image only.
AEMC Mfr # 8220 W/MN193-BK – Item # EW-20019-82
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionPower Quality Meter with 6A/120Aac Probe Model 8220 with MN93-Bk
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
FEATURES
  • Measures up to 600VRms or VDC
  • Measures up to 6500AAC or 1400ADC (probe dependent)
  • Displays Min, Max and Average volts and amps, crest factor, peak value and K-factor
  • Calculates and displays watts, vARs and VA, power factor and displacement power factor or single-phase and balanced three-phase
  • Displays total harmonic distortion (THD-F and THD-R) for voltage and current
  • Displays individual harmonic values and % for volts and amps from DC through the 50th harmonic
  • Captures and displays inrush current
  • Calculates and displays phase rotation and RPM
  • Displays temperature in both °F and °C
  • Displays resistance up to 200O
  • Conducts continuity and diode tests
  • Stores up to nine complete sets of readings for all volt, amp, power and harmonic measurements
  • Configurable from DataView® software via optically coupled RS232 port
  • Operates off batteries or recharger
Standard accessories include: current probes with 10 ft leads and black connector, black and red 10 ft voltage leads and alligator clips, RS232 optically coupled serial cable, NiMH battery, US 120V power cord, DataView® software, carrying bag, soft carrying pouch and user manual.