AEMC CA7024 Fault Mapper Alpha Numeric TDR
Representative image only.
AEMC Mfr # 2127.8 – Item # EW-17782-01
  • Fault Mapper
    Cable Length Meter and Fault Locator Alphanumeric TDR - AE212780
  • Handheld cable length meter and fault locator
  • Detects opens and shorts and
  • the distance to them from one end of the cable
  • Measure cable length up to 6000 ft or 2000m (selectable)
  • Automatic cable impedance compensation
  • Built-in library of most common cables and their Velocity of Propagation (Vp)
  • Manual selection of Vp for all cables not found in library
  • User programmable cable library with 15 positions
  • Vp setting displayed along with length and cable type (if in library)
  • Built-in tone generator for tracing and locating cables
  • Large high-visibility blue electroluminescent backlit display
  • 1 year warranty
  • SPECS & DESCRIPTION
    Specifications & Description
    • DescriptionFault Mapper Alpha Numeric TDR
    • Warranty1 Year
    1 Year Warranty
    Key Features
    • Fault Mapper
      Cable Length Meter and Fault Locator Alphanumeric TDR - AE212780
    • Handheld cable length meter and fault locator
    • Detects opens and shorts and
    • the distance to them from one end of the cable
    • Measure cable length up to 6000 ft or 2000m (selectable)
    • Automatic cable impedance compensation
    • Built-in library of most common cables and their Velocity of Propagation (Vp)
    • Manual selection of Vp for all cables not found in library
    • User programmable cable library with 15 positions
    • Vp setting displayed along with length and cable type (if in library)
    • Built-in tone generator for tracing and locating cables
    • Large high-visibility blue electroluminescent backlit display
  • 1 year warranty
  • More About this Item
    Wire Mapper Pro LAN Cable Tester, Fault Mapper Cable Length Meter and Fault Locator Alphanumeric TDR, and Fault Mapper Pro Telephone / Coaxial / Parallel Cable Tester