AEMC CA7028 Wire Mapper Pro Cable Tester
Representative image only.
AEMC Mfr # 2127.82 – Item # EW-17782-03
  • Fault Mapper
    Cable Length Meter and Fault Locator Alphanumeric TDR - AE212780
  • Handheld cable length meter and fault locator
  • Detects opens and shorts and
  • the distance to them from one end of the cable
  • Measure cable length up to 6000 ft or 2000m (selectable)
$749.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionWire Mapper Pro Cable Tester
Key Features
  • Fault Mapper
    Cable Length Meter and Fault Locator Alphanumeric TDR - AE212780
  • Handheld cable length meter and fault locator
  • Detects opens and shorts and
  • the distance to them from one end of the cable
  • Measure cable length up to 6000 ft or 2000m (selectable)
  • Automatic cable impedance compensation
  • Built-in library of most common cables and their Velocity of Propagation (Vp)
  • Manual selection of Vp for all cables not found in library
  • User programmable cable library with 15 positions
  • Vp setting displayed along with length and cable type (if in library)
  • Built-in tone generator for tracing and locating cables
  • Large high-visibility blue electroluminescent backlit display
More About this Item
Wire Mapper Pro LAN Cable Tester, Fault Mapper Cable Length Meter and Fault Locator Alphanumeric TDR, and Fault Mapper Pro Telephone / Coaxial / Parallel Cable Tester
Call Us At1-847-549-7600
Live chat