This item is now discontinued. Please browse similar items
Afab In-Line Process Refractometer, 1.3 to 1.49 Refractive Index; 240 VAC (Reconditioned)
Representative image only.
Afab Mfr # 81005-05 RECONDITIONED – Item # EW-00248-GD
  • Send sata to chart recorder or control processing equipment in real time using 4 to 20 mA output
  • 0 to 100% analog display meter is easily correlated to your specific monitoring needs: Be, Brix, specific gravity, etc.
SPECS & DESCRIPTION
Specifications & Description
Includes
standard calibration for 0 to 40 Brix and 80 to 100°F, a 10-ft power cable, and a 20-ft interconnecting cable. An adapter is required to mount the sensor into your system — order separately.
Key Features
  • Send sata to chart recorder or control processing equipment in real time using 4 to 20 mA output
  • 0 to 100% analog display meter is easily correlated to your specific monitoring needs: Be, Brix, specific gravity, etc.
More About this Item
Monitor and control the concentration of dissolved solids or water soluble liquids in your process lines with this in-line process refractometer. Refractometer accurately measures the refractive index of your process, providing temperature compensated readings that can easily be correlated to a percent solids, Brix, Be, or specific gravity scale.
Easy user-controlled required calibration and temperature compensation capability allows you to tailor calibration and compensation to your specific process. Refractometer is factory-calibrated to 0 to 40% Brix in an operating range of 80 to 100°F. Required custom calibration (81005-20) is available within a measuring range of 0 to 100% Brix equivalent. Temperature compensate for spans of ±20°F over a range of 0 to 250°.
Sensing unit features state-of-the-art, silicone detector with stainless steel sensing head and sapphire prism that remain inert to essentially all liquids over teh operating temperature range of the device. High-intensity visible LED light source is designed for long life and low maintenance. Built-in AGC loop compensates for LED aging and suspended solids, guaranteeing maximum accuracy.
NEMA 4 head and console are watertight and drip proof. Refractometer operates on 120 or 240 VAC input power - no special wiring required. Unit can be mounted onto the side of a tank or can be installed in-line.