Air Impurities Removal Systems RF-400-1 Secondary Merv 8 Pleated Filter for Extract-All Systems
Representative image only.
Air Impurities Removal Systems Mfr # RF-400-1 – Item # EW-33704-90
$133.25USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionSecondary Merv 8 Pleated Filter for Extract-All Systems
More About this Item
High-Efficiency Filters capture smoke and dust — ideal if you mix dry chemicals, sand, grind, or do light welding or soldering. Prefilter is 30 to 35% efficient; high-efficiency cartridge is 96.5% efficient.
HEPA Filters capture soldering fumes and vapors that contain lead. Prefilter is 30 to 35% efficient; HEPA filter is 99.97% efficient.
Final Adsorption Carbon Module captures volatile solvents, touch-up paint fumes, and other noxious odors. Available in combination with primary high-efficiency or HEPA filter (33703-02 and -04), or use asa final stage for portable air cleaning system.