Air Impurities Removal Systems Activated Carbon Refill for Ductless Air Filter Systems, fits 33703-45
Representative image only.
Air Impurities Removal Systems Mfr # RF-987-4A – Item # EW-33703-47
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionActivated Carbon Refill for Ductless Air Filter Systems, fits 33703-45
More About this Item
Easily install or change filters in just seconds. Filter packages designed to suit your specific working conditions
High-Efficiency Filters captures smoke and dust — ideal if you mix dry chemicals, sand, grind, or do light welding. If you use solvents or paint in the same process, choose a combination high-efficiency filter with refillable 2" thick carbon module. Filter is 95% efficient.
HEPA Filters capture soldering fumes and vapors that contain lead. If you use solvents or paint in the same process, choose a combination HEPA filter with refillable 2" thick carbon module. Filter is 99.97% efficient.
Activated Carbon Filter captures volatile solvents, touch-up paint fumes, and other noxious odors. The primary