Allegro 4001 Respirator Cleaning Kit, includes 90 soap packs and 100 sealable bags
Representative image only.
Allegro Mfr # 4001 – Item # EW-86332-20
Meets OSHA recommendation 29 CFR 1910.134 Appendix B
  • Two-sided bucket allows you to wash and rinse respirators in the same bucket
$140.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionRespirator Cleaning Kit, includes 90 soap packs and 100 sealable bags
Includes
90 soap packets, 17-qt (16-L) two-sided bucket, air bulb, scrubbing brush, and 100 storage bags with seals.
Key Features
  • Two-sided bucket allows you to wash and rinse respirators in the same bucket
More About this Item
Keep your half- and full-facepiece respirators clean while complying with OSHA recommendations. Kit includes a water-soluble detergent that won''t harm sensitive rubber components. Concentrated soap is prepackaged in 1/2 oz. (14.7 mL) packets for just the right amount every time.