Almatec FUTUR 20 AODD, max. 100-200°C High Temperature PTFE Housing, PTFE Diaphragms, PTFE Valve Balls, 1/2" NPT
Representative image only.
Almatec Mfr # Fut20H – Item # EW-76980-08
German precision engineered AODD pumps to get your application up and flowing.
PRODUCT OPTIONS
$5,421.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionAODD Pump, Max. 100-200°C High Temperature PTFE Housing, PTFE Diaphragms, PTFE Valve Balls, 1/2" NPT
More About this Item
The heart of the AODD is the air-valve and thoughtful German engineering went into the development of the precision control PERSWING P® air valve and its Air Distribution System (ADS). Resulting in a metal-free, pneumatically pilot-operated PERSWING P® (ADS) that ensures accurate reversal of the main piston and offers low noise levels, minimal maintenance and lube-free operation (due to two moving parts).
Almatec plastic pumps feature a solid body construction made out of virgin grade PE or PTFE materials that support the forces generated within a reciprocating pump. The high static mass leads to maximum process fluid containment and eliminates the need for external metal parts that would otherwise be used. The mechanical machining of a solid plastic block is economical thanks to modern CNC technology, enabling tight tolerances and maximum value. Almatec also feature best-in-class diaphragms that provide a smooth profile that is uninterrupted by any seals, maximizing containment and clean-ability. The diaphragms are designed to simplify assembly, reduce air consumption and optimize performance.