Ametek 831TG1000BLS 0-1000 Psig Fixed Range Press Transmitter
Representative image only.
Ametek Mfr # 831TG1000BLS – Item # EW-68448-80
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Min Pressure (PSI)0
  • Max Pressure (PSI)1000
  • Description0-1000 Psig Fixed Range Press Transmitter
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
FEATURES
  • A miniature, low-priced fixed range transmitter, just 0.83 lbs. (374 grams)
  • All welded 316 stainless steel construction and wetted parts (no aluminum)
  • Ranges form vacuum to 5000 psi (0 to 1 to 0 to 345 bar)
    ±0.30% accuracy
  • designed to meet FM Explosion-proof approvals (Consults factory for approvals)
  • 4 to 20 mA output at 12 to 30 VDC
  • 3/4-Inch NPT Female conduit connection cable
  • 1/2-Inch NPT Female pressure port connection
  • Straight in-line construction for a slim profile
  • 24 Inch cable standard