Amphenol 8007 CO2 Sensor with Display, Duct Mount
Representative image only.
Amphenol Mfr # T8300-DB – Item # EW-10101-47
$329.00USD / Each
$176.00USD / Each
for an additional
$176.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionCO2 Sensor with Display, Duct Mount
More About this Item
FEATURES
  • Gas sensing engine provides high accuracy in a compact low cost package
  • ABC Logic™ self-calibration system eliminates the need for manual calibration in most applications
  • Gold plated optical sensor increases sensor life and durability
  • Mounting bracket with terminal block provides quick, easy wiring
  • Dual simultaneous analog output (V & mA)
  • On board relay with adjustable set point and dead band
  • Choice of 9 preprogrammed ''standard settings'' on easy to use interface
  • PC interface and adjustable settings allows for simple configuraiton
  • Optional enclosures available
  • Lifetime calibration guarantee