Amprobe CR50A Capacitance and Resistance Meter from
Representative image only.
Amprobe Mfr # CR50A – Item # EW-20021-35
Available for sale only in the United States and Canada
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Capacitance Range0.1pF in 200pF
  • Capacitance Resolution20,000µF
  • Resistance Range0.01 ohm in 20 ohm
  • Resistance Resolution20M ohm
  • Diode TestYes
  • DescriptionCapacitance and Resistance Meter from
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
The MNCR50 is a pocket-sized capacitance and resistance meter that measures capacitance in 9 ranges from 200pF to 20mF; seven ranges for resistance from 20 ohm to 20Mohm. The MNLCR55 measures inductance from 200uH to 200H; capacitance measures ranges from 200pF to 20mF, and resistance from 20ohm to 20Mohm.