Amprobe DC205C Deluxe Hard Shell Carrying Case
Representative image only.
Amprobe Mfr # DC205C – Item # EW-20008-11
Built to be tough, reliable, safe and easy to use
  • Highest voltage capability, measuring to 1500 VDC and 1000 VAC
  • IP67 rated with special sealing for environment and water resistance
  • Constructed with drop-resistant housing and PCB mounting
  • CAT IV 1000 V rated
  • Auto power-down after 30 minutes of inactivity
  • Low battery indication
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionDeluxe Hard Shell Carrying Case
  • Warranty1 year
1 Year Warranty
Key Features
  • Highest voltage capability, measuring to 1500 VDC and 1000 VAC
  • IP67 rated with special sealing for environment and water resistance
  • Constructed with drop-resistant housing and PCB mounting
  • CAT IV 1000 V rated
  • Auto power-down after 30 minutes of inactivity
  • Low battery indication
  • 1 year warranty
More About this Item
Designed for professional use in harsh environments, the heavy duty multimeters feature waterproof construction to prevent dirt, grime and moisture from entering the case and causing inaccurate readings or damaging shorts. Built of tough, fire-retardant thermoplastic, the multimeters are resistant to grease and chemical spills, with all sensitive components shock-mounted for a drop resistant to at least eight feet.
The extended voltage measurement capability of 1500 VDC and 1000 VAC is unmatched by any other line of digital multimeters. The extended voltage ranges can withstand transients up to 12 kV.
Available for sale only in the United States and Canada.