Amprobe DL248D Deluxe Test Lead Kit [AMP]
Representative image only.
Amprobe Mfr # DL248D – Item # EW-23031-64
Available for sale only in the United States and Canada.
  • Silicon-insulated
  • high quality test lead wires - one end has shrouded straight banana plug, other end with a shrouded right-angle banana plug
  • One red and one black lead, 48 in. long
  • Maximum voltage rating 1000VDC or AC peak
  • IEC 1010-2 approved
SPECS & DESCRIPTION
Specifications & Description
  • DC Max (Volts)1500
  • AC Max (Volts)1500
  • DescriptionDeluxe Test Lead Kit [AMP]
Key Features
  • Silicon-insulated
  • high quality test lead wires - one end has shrouded straight banana plug, other end with a shrouded right-angle banana plug
  • One red and one black lead, 48 in. long
  • Maximum voltage rating 1000VDC or AC peak
  • IEC 1010-2 approved
More About this Item
Deluxe Test Lead Kit DL248D Includes:
- Test Probes with 1/2'' tip length (1 red, 1 black)
- Insulated alligator clips (1 red, 1 black)
- Spade lugs (1 red, 1 black)
- Crimp style spring hooks (1 red, 1 black)
- 2'' long Probe extender (1 only red)
- Rugged cordura case with storage pockets - top pocket has hook-and-loop closure to prevent loss of small items, folds to convenient pocket size