Amprobe ELS2A AC Current Clamp Accessory
Representative image only.
Amprobe Mfr # ELS2A – Item # EW-20029-42
Measures current without splitting AC line cords
  • Includes x10 multiplier loop to accurately measure low-current
  • Check for voltage sags and surges with built-in voltage measurement points
  • Rated for 120 VAC, 15 Amps
$26.99USD / Each
$29.99USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionAC Current Clamp Accessory
Key Features
  • Includes x10 multiplier loop to accurately measure low-current
  • Check for voltage sags and surges with built-in voltage measurement points
  • Rated for 120 VAC, 15 Amps
More About this Item
Measures current without splitting AC line cords.
Available for sale in the United States and Canada only