Amprobe INSP-3 Wiring Inspection / Circuit Tester
Representative image only.
Amprobe Mfr # INSP-3 – Item # EW-20037-37
Rugged tester is designed to verify building wiring compliance to electrical codes Available for sale in the United States and Canada only
  • Single large display saves operator time - no scrolling or switching screens needed
  • Meter detects faulty wiring in need of repair without removing outlets, cover plates, or panel covers
  • Identify faulty splices and connections, incorrect wiring, undersized wiring, faulty GFCIs, faulty or incorrectly wired AFCI’s, incorrect line voltage, and poor ground quality
  • User selectable 0, 10, 15, and 20 amp loads to verify performance of the electrical system
  • Incorrect wiring or voltage drop test failure is clearly indicated by flashing screen
  • Will not trip circuit breakers or blow fuses during the test
  • Tests GFCI and AFCI operations
  • Check ground quality for safety and ability to support sensitive electronic equipment
  • Measure fault currents
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionWiring Inspection / Circuit Tester
  • Warranty1 Year
1 Year Warranty
Key Features
  • Single large display saves operator time - no scrolling or switching screens needed
  • Meter detects faulty wiring in need of repair without removing outlets, cover plates, or panel covers
  • Identify faulty splices and connections, incorrect wiring, undersized wiring, faulty GFCIs, faulty or incorrectly wired AFCI’s, incorrect line voltage, and poor ground quality
  • User selectable 0, 10, 15, and 20 amp loads to verify performance of the electrical system
  • Incorrect wiring or voltage drop test failure is clearly indicated by flashing screen
  • Will not trip circuit breakers or blow fuses during the test
  • Tests GFCI and AFCI operations
  • Check ground quality for safety and ability to support sensitive electronic equipment
  • Measure fault currents
  • 1 year warranty
More About this Item
This wiring inspector saves you valuable time and money by eliminating guess work. Easily verify if wiring is tested for load carrying ability and if it meets electrical code recommendations for voltage drop under load. Unit provides testing efficiency of relevant test data - voltage, voltage drop, hot and neutral voltage drop, voltage with load, and ground impedance.