Amprobe TL1500DC Test Lead Set
Representative image only.
Amprobe Mfr # TL1500DC – Item # EW-20051-24
Silicone insulation protects hands from shock and heat
  • Flexible silicone insulated leads are heat and cold resistant
  • Alligator clips provide a secure, hands-free connection
  • Spade lugs clamp to conductors with compression nuts or screws
  • 1 year warranty
$37.03USD / Each
$38.99USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Jaw Size0.3
  • DescriptionTest Lead Set
  • Warranty1 year
1 Year Warranty
Includes
Two lead wires, two standard probe tips, two alligator clips, and two spade lugs.
Key Features
  • Flexible silicone insulated leads are heat and cold resistant
  • Alligator clips provide a secure, hands-free connection
  • Spade lugs clamp to conductors with compression nuts or screws
  • 1 year warranty
More About this Item
This multipurpose test lead set contains two 48" long silicone-insulated lead wires, two standard probe tips, two alligator clips, and two spade lugs for use in various electrical testing applications. The silicone insulation on the lead wires resists melting in hot environments as well as becoming brittle in cold temperatures. Use lead wires with testing devices that have universal 4 mm jacks. The alligator clips with teeth grip the conductor securely for hands-free operation; the clips are 3.15" long and have 0.30" jaw opening.