Ansell TouchNTuff® 69-210 Latex Gloves, Small, 100/bx
Representative image only.
Ansell Mfr # 69-210-516704 – Item # EW-86568-00
Reliable barrier protection with maximum degree of dexterity
  • Tear resistance
  • Ideal for general laboratory
PRODUCT OPTIONS
$17.70USD / Box of 100
BUY MORE & SAVE
  • 1-9$17.70USD / Box of 100
  • 10+$15.93USD / Box of 100
SPECS & DESCRIPTION
Specifications & Description
  • Thickness (mil)5
  • MaterialLatex
  • SizeSmall
  • Length (in)9 1/2
  • Length (cm)24
  • ColorWhite
  • CuffBeaded
  • SterileNo
  • DescriptionLatex Gloves, small, 100/bx
Key Features
  • Tear resistance
  • Ideal for general laboratory
More About this Item
Natural rubber latex gloves are 3.5-mil thick for strength and tear-resistance. Inner polymer coating for easy donning and doffing. General purpose gloves are ideal for general laboratory, parts handling, electronics assembly, or quality control applications. Sold in box of 100.