Ansell Microflex® V28 Light Duty Disposable Vinyl Glove, Size M (7.5-8); 100/Box
Representative image only.
Ansell Mfr # 163297 BOX – Item # EW-81610-49
Cost-effective protection for light duty tasks
  • Free from natural rubber latex
  • Produced without use of DEHP
  • Ambidextrous
PRODUCT OPTIONS
$11.20USD / Box of 100
SPECS & DESCRIPTION
Specifications & Description
  • Thickness (mil)2.8
  • MaterialVinyl
  • SizeMedium
  • Length (in)9 1/2
  • Length (cm)24
  • ColorClear
  • CuffStandard
  • ComplianceFDA 21 CFR 177-2602
  • SterileNo
  • PowderedNo
  • DescriptionLight Duty Disposable Vinyl Glove, Size M (7.5-8); 100/Box
Key Features
  • Free from natural rubber latex
  • Produced without use of DEHP
  • Ambidextrous
More About this Item
These disposable vinyl gloves are ideal for food handling, light-duty cleaning, and general-purpose use. The gloves offer a smooth finish, giving you a more natural fit. Powder-free to reduce skin irritation. Disposable—for single use only.