Ansell Microflex® V28 Light Duty Disposable Vinyl Gloves, Size M (7.5-8); 1000/CS (10 boxes of 100 gloves)
Representative image only.
Ansell Mfr # 163297 – Item # EW-81620-74
Cost-effective protection for light duty tasks
  • Free from natural rubber latex
  • Produced without use of DEHP
  • Ambidextrous
PRODUCT OPTIONS
$94.00USD / Case of 1000
SPECS & DESCRIPTION
Specifications & Description
  • MaterialVinyl
  • Thickness (mil)2.8
  • SizeMedium
  • Length (in)9 1/2
  • Length (cm)24
  • ColorClear
  • CuffStandard
  • ComplianceFDA 21 CFR 177-2600
  • SterileNo
  • PowderedNo
  • DescriptionLight Duty Disposable Vinyl Gloves, Size M (7.5-8.0)
Key Features
  • Free from natural rubber latex
  • Produced without use of DEHP
  • Ambidextrous
More About this Item
These disposable vinyl gloves are ideal for food handling, light-duty cleaning, and general-purpose use. The gloves offer a smooth finish, giving you a more natural fit. Powder-free to reduce skin irritation. Disposable—for single use only.