Ansell Microflex® V29 Blue Disposable Vinyl Glove, Size L (8.5-9); 100/Box
Representative image only.
Ansell Mfr # 163310 BOX – Item # EW-81610-54
Cost-effective protection for light duty tasks
  • Free from natural rubber latex
  • Produced without use of DEHP
  • Ambidextrous
PRODUCT OPTIONS
$7.00USD / Box of 100
SPECS & DESCRIPTION
Specifications & Description
  • MaterialVinyl
  • Thickness (mil)2.8
  • SizeLarge
  • Length (in)9 1/2
  • Length (cm)24
  • ColorBlue
  • CuffStandard
  • ComplianceFDA 21 CFR 177-2600
  • SterileNo
  • PowderedNo
  • DescriptionBlue Disposable Vinyl Glove, Size L (8.5-9); 100/Box
Key Features
  • Free from natural rubber latex
  • Produced without use of DEHP
  • Ambidextrous
More About this Item
These disposable blue vinyl gloves provide outstanding elasticity and tensile strength, ideal for food preparation, light-duty cleaning, and general maintenance. The gloves offer a smooth finish, giving you a more natural fit. Powder-free to reduce skin irritation. Disposable—for single use only.