Ansell TouchNTuff 92-500 Nitrile Gloves, Powdered, Small
Representative image only.
Ansell Mfr # 92-500 585188 – Item # EW-81602-40
Standard in comfort, durability, and cleanliness
  • Resist punctures and tear
  • Ambidextrous
PRODUCT OPTIONS
$25.30USD / Box of 100
BUY MORE & SAVE
  • 1-9$25.30USD / Box of 100
  • 10+$22.77USD / Box of 100
SPECS & DESCRIPTION
Specifications & Description
  • Thickness (mil)4
  • MaterialNitrile
  • SizeSmall
  • Length (in)9 1/2
  • Length (cm)24
  • ColorGreen
  • ComplianceFDA-accepted
  • SterileNo
  • PowderedYes
  • DescriptionNitrile Gloves, Powdered, Small
Key Features
  • Resist punctures and tear
  • Ambidextrous
More About this Item
Premium quality gloves are thin enough to allow excellent dexterity, yet tough enough to resist punctures and tears. Gloves are 100% nitrile and contain no fillers, silicones, or plasticizers.