Ansell TouchNTuff® 92-616 Lightweight Disposable Nitrile Glove, S (6, 5-7); 150/Box
Representative image only.
Ansell Mfr # 105131 BOX – Item # EW-81610-44
Puncture and chemical splash resistant
  • Compliant with food handling requirements
  • Packaged 150 gloves per dispenser
  • Ambidextrous
PRODUCT OPTIONS
$29.90USD / Box of 150
SPECS & DESCRIPTION
Specifications & Description
  • MaterialNitrile
  • Thickness (mil)3.1
  • SizeSmall
  • Length (in)9 1/2
  • Length (cm)24
  • ColorBlue
  • GripTextured fingertips
  • CuffStandard
  • ComplianceFDA 21 CFR 177-2600
  • SterileNo
  • PowderedNo
  • DescriptionLightweight Disposable Nitrile Glove, S (6, 5-7); 150/Box
Key Features
  • Compliant with food handling requirements
  • Packaged 150 gloves per dispenser
  • Ambidextrous
More About this Item
These lightweight nitrile gloves provide you comfort and durability. Made of 100% nitrile to prevent Type 1 allergies. These gloves offer you puncture and chemical splash protection from hydrocarbons, fats, grease, and alcohols. Ideal for food processing applications. Packaged 150 gloves per dispenser to minimize packaging.Ten packages per case.