Ansell TouchNTuff® 93-250 Grey Disposable Nitrile Glove, Size M (7, 5-8); 100/Box
Representative image only.
Ansell Mfr # 118248 BOX – Item # EW-81610-57
Maintain a secure grip on wet, oily, and dry objects
  • Soft nitrile formulation provides hours of comfort
  • Free from natural rubber latex
  • Ambidextrous
SPECS & DESCRIPTION
Specifications & Description
  • MaterialNitrile
  • Thickness (mil)5.1
  • SizeMedium
  • Length (in)9 19/32
  • Length (cm)24.5
  • ColorGrey
  • GripTextured fingers
  • CuffStandard
  • ComplianceFDA 21 CFR 177-2600
  • SterileNo
  • PowderedNo
  • DescriptionGrey Disposable Nitrile Glove, Size M (7, 5-8); 100/Box
Key Features
  • Soft nitrile formulation provides hours of comfort
  • Free from natural rubber latex
  • Ambidextrous
More About this Item
These dark grey nitrile gloves decrease fatigue and increase productivity by reducing the amount of force needed to grip wet, oily, and dry objects. The gloves soft, robust, nitrile formulation offers enhanced chemical splash protection from a wide range of acids, bases, and solvents. Silicone free design is paint and finish process friendly. Disposable—for single use only.