Antimicrobial Pleated Polyester Filter Cartridge; 9.625" L, 20 um (nominal)
Representative image only.
Item # EW-29808-41
$116.60USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Filter MediaPolyester
  • Micron Rating20
  • Filter Length (in)9.625
  • DescriptionAntimicrobial Pleated Polyester Filter Cartridge; 9.625" L, 20 um (nominal)
More About this Item
Antimicrobial Cartridges are pleated polyester with interwoven silver zeolite fibers. The interwoven fibers ensure long-lasting protection against algae, odor-causing bacteria, mold, mildew, and biofilms that might otherwise grow on the cartridge surface.