Anton Paar 88085 Infrared to USB Adapter for Digital Density / Specific Gravity Meters
Representative image only.
Anton Paar Mfr # 88085 – Item # EW-25750-61
Compact design and wide grip for easy one-hand operation — perfect for on-site inspections
PRODUCT OPTIONS
$70.45USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionInfrared to USB Adapter for Digital Density/Specific Gravity Meters
Includes
Carrying case
More About this Item
This density meter is easy to grip and control even when you're wearing thick work gloves. The narrow 1" (2.5 cm) depth lets you maneuver in tight spaces. Meter features a pipette-style pump for good sample flow control — no bellows or bulb to rupture and leak. To draw in a sample, simply press the lever at the top of the meter with your thumb. View density, specific gravity, % concentration, or sample temperature on the large LCD. Press lever again to discharge sample.
Store up to 1024 readings with a push of a button. Access permanently stored concentration tables including % Brix, % alcohol, proof, °Baume, °Plato, API gravity, specific gravity at reference temperature, and % H2SO4. Durable meter housing is sealed against pump spills. Infrared interface lets you send stored readings to your PC for later analysis — order infrared to USB adaptor or optional infrared printer separately. Case is included.