Applikon BioBundle Z001023551 pH Sensor for Fermentation System, 235 mm
Representative image only.
Applikon BioBundle Mfr # Z001023551 – Item # EW-29207-22
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionpH Sensor for Fermentation System, 235 mm
More About this Item
These pH sensors use a nontoxic gel to prevent silver/protein interference on the diaphragm. Each sensor has a Ti layer on the innertube to ensure no window between sensor head and gel which results in an excellent screen to protect against electromagnetic interference. Pre-autoclaved; MSDS and GLP certificate available upon request.