Applikon Chemostat Tube for 15-Liter Fermentation/Bioreactor Systems
Representative image only.
Applikon Mfr # Z811310005 – Item # EW-01919-46
$1,394.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionChemostat Tube for 15-Liter Fermentation/Bioreactor Systems
More About this Item
Chemostat tubes are used with a Masterflex® tubing pump (sold separately) to draw off harvested media or to add fresh media to maintain constant media level.