Applikon Z365001296 MicroFlask Standard Deep-Well Sandwich Cover, 96-Well
Representative image only.
Applikon Mfr # Z365001296 – Item # EW-29208-11
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionMicroFlask Standard Deep-Well Sandwich Cover, 96-Well
More About this Item
Sandwich Covers with Layers
These sandwich covers have re-usable layers. They include a stainless steel lid for rigid support, a microfiber cotton layer, ePTFE (0.3 µm pores) layer for sterility, and a soft silicone layer to seal the system. All components can handle repeated autoclaving.