Arcis Sample Prep Kit, 50 reactions
Representative image only.
Arcis Mfr # UFL002B – Item # EW-13204-52
Fast and convenient sample prep in minutes
  • Protocol requires only 2 steps and takes less than 3 minutes
  • Compatible with a wide variety of downstream applications
  • No heating or centrifugation steps required
  • Stabilizes DNA for up to 30 days at ambient temperatures
  • 18 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Test TypeDNA
  • Number of Tests Included50
  • DescriptionSample Prep Kit, 50 rxn
  • Warranty18 months
18 Month Warranty
Key Features
  • Protocol requires only 2 steps and takes less than 3 minutes
  • Compatible with a wide variety of downstream applications
  • No heating or centrifugation steps required
  • Stabilizes DNA for up to 30 days at ambient temperatures
  • 18 month warranty
More About this Item
Quick and easy sample preparation kit provides rapid amplification of genomic DNA from a variety of samples. This complete, ready-to-use kit lets you to go from sample to amplifiable nucleic acids in under 3 minutes. There is no need for any prior sample preparation or additional isolation or purification.
The sample is prepared and stabilized without need for heating or centrifugation steps. The kit is suitable for use in PCR and qPCR reactions using standard and fast cycling protocols. The product can be used on virtually any DNA containing sample. The nucleic acid released has been successfully applied to molecular biology techniques including PCR, sequencing, and cloning.