Argos Technologies CapLock™ Microcentrifuge Tubes, 2.0 mL, 500/PK

Representative image only.
Argos Technologies Item # EW-24410-79
Ideal for applications where sample integrity is importance
  • Free of RNase, DNase, endotoxins, and human DNA
  • Certified TSE/BSE free
  • Withstands centrifugation up to 30,000 g

PRODUCT OPTIONS

$24.18USD / Pkg of 500
$32.25USD / Pkg of 500

SPECS & DESCRIPTION

Specifications & Description

  • Max RCF x g30000
  • SterileNo
  • AutoclavableYes
  • Tube Capacity (mL)2
  • Tube MaterialPolypropylene
  • Tube ColorClear
  • Tube Bottom ShapeRound
  • Cap StyleDouble-lock
  • GraduationsYes
  • Packaging500/Pk
  • Special FeaturesCapLock™ cap closing system
  • DescriptionCapLock Microcentrifuge Tubes, 2.0 mL, 500/PK

Key Features

  • Free of RNase, DNase, endotoxins, and human DNA
  • Certified TSE/BSE free
  • Withstands centrifugation up to 30,000 g

More About this Item

CapLock™ microcentrifuge tubes features a double-lock cap closing system to ensure samples are safe and secure. It is ideal for boiling applications and working with DNA or hazardous substances. The polypropylene (PP) tube is autoclavable at 250°F (121°C) and freezable to -148°F (-100°C). Other features include an extended lip for easy opening, marked graduations, and frosted areas on tube and cap.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat