Cole-Parmer Essentials L-Shaped Cell Spreaders, Sterile, Polypropylene, Individually Wrapped; 100/Cs
Representative image only.
Cole-Parmer Essentials Item # EW-04396-52
Spread bacterial cultures evenly over agar in petri dishes
  • Sterile to prevent contamination risk
  • L-shape allows application over the entire dish or plate surface
  • Curved end protects agar
PRODUCT OPTIONS
$34.00USD / Case of 100
SPECS & DESCRIPTION
Specifications & Description
  • SterileYes
  • Overall Length (mm)145
  • Overall Length (in)5 45/64
  • ColorBlue
  • MaterialPolypropylene
  • DescriptionL-Shaped Cell Spreaders, Sterile, Polypropylene, Individually Wrapped; 100/Cs
Sterile
Key Features
  • Sterile to prevent contamination risk
  • L-shape allows application over the entire dish or plate surface
  • Curved end protects agar
More About this Item
Apply cultures over an entire petri dish or plate with these sterile polypropylene cell spreaders. This L-shaped tool has a smooth spreading surface with a slight upward return to reduce the risk of damaging the agar.
Manufactured under ISO 13485 and ISO 9001 quality management systems in an ISO Class 8 (formerly Class 100,000) cleanroom environment, these cell spreaders are radiation sterilized (SAL 10-6), DNase/RNase free, nonpyrogenic, and noncytotoxic. Available individually wrapped or bulk packed.