Argos Technologies Disposable filter upper cup, PES membrane, 0.45um, 250 mL, 24/CS
Representative image only.
Argos Technologies Item # EW-07630-92
Save time with presterilized disposable filtration units
  • Individually wrapped for sterility
  • Easy-pouring large cap on reservoir
  • Graduations on funnel and reservoir for improved accuracy
  • Hose connector fits multiple hose diameters
PRODUCT OPTIONS
$93.00USD / Case of 24
$124.00USD / Case of 24
SPECS & DESCRIPTION
Specifications & Description
  • Membrane MaterialPolyethersulfone (PES)
  • Pore Size (µm)0.45
  • Housing MaterialPolypropylene (PP)
  • Capacity (mL)250
  • SterileYes
  • ConnectionsHose Barb
  • DescriptionDisposable filter upper cup, PES membrane, 0.45um, 250 mL, 24/CS
Key Features
  • Individually wrapped for sterility
  • Easy-pouring large cap on reservoir
  • Graduations on funnel and reservoir for improved accuracy
  • Hose connector fits multiple hose diameters
More About this Item
Disposable bottletop aspirators provide a complete vacuum filtration solution for your separation and purification needs. All units come presterilized by gamma irradiation and certified nonpyrogenic, and are individually wrapped in easy-to-open packages. The large openings on the reservoir and funnel make for easy access. The funnel is made of clear polystyrene for easy viewing, and the bottle and filter adapter are made of 100% virgin polypropylene.