Argos Technologies FlexiFuge™ Fixed-Angle Rotor for Flexifuge, 4 x 5.0 mL
Representative image only.
Argos Technologies Item # EW-22999-55
Versatile rotor options suit many standard lab needs
  • Quickly and easily interchange rotors
  • Constructed of durable polycarbonate
  • Microtube 4 x 5 mL rotor
PRODUCT OPTIONS
$56.75USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Rotor TypeFixed-angle
  • Tube Sizes Accepted5 mL
  • Number Of Tubes Accepted4
  • DescriptionFixed-AngleRotorforFlexifuge,4x5.0mL
Key Features
  • Quickly and easily interchange rotors
  • Constructed of durable polycarbonate
  • Microtube 4 x 5 mL rotor
More About this Item
Replacement rotors and for the Argos Technologies® Flexifuge Centrifuge, models 22999-50 through -52.
This rotor is suitable for a relative centrifugal force of 2414 G at 6000 rpm and 5677 G at 9200 rpm.