Argos Technologies FlexiFuge™ Rotor for Flexifuge 16 x Standard PCR tubes
Representative image only.
Argos Technologies Item # EW-22999-54
Versatile rotor options suit many standard lab needs
  • Quickly and easily interchange rotors
  • Constructed of durable polycarbonate
  • Strip PCR 8 x 0.2 mL tube strips or 16 x 0.2 mL tube rotor
PRODUCT OPTIONS
$56.25USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Rotor TypeFixed-angle
  • Tube Sizes AcceptedStandard PCR tubes
  • Number Of Tubes Accepted16
  • DescriptionRotorforFlexifuge16xStandardPCRtubes
Key Features
  • Quickly and easily interchange rotors
  • Constructed of durable polycarbonate
  • Strip PCR 8 x 0.2 mL tube strips or 16 x 0.2 mL tube rotor
More About this Item
Replacement rotors and for the Argos Technologies® Flexifuge Centrifuge, models 22999-50 through -52.
This rotor is suitable for a relative centrifugal force of 1509 G at 6000 rpm and 3549 G at 9200 rpm.