Argos Technologies LiteSafe® 0.5 mL Black LiteSafe Microcentrifuge Tubes - 500/pk
Representative image only.
Argos Technologies Item # EW-06333-75
Protect photosensitive samples
  • Unlike amber tinted tubes, these are completely opaque protecting even the most sensitive samples
  • RNase and DNase free
PRODUCT OPTIONS
$22.01USD / Pkg of 500
$29.35USD / Pkg of 500
SPECS & DESCRIPTION
Specifications & Description
  • Max RCF x g15000
  • Tube Capacity (mL)0.5
  • Tube MaterialPolypropylene
  • Tube ColorBlack
  • Tube Bottom ShapeConical
  • Cap IncludedYes
  • Cap StyleClick-Fit
  • Cap MaterialPolypropylene
  • GraduationsNone
  • Description0.5 mL Black LiteSafe Microcentrifuge Tubes - 500/pk
Key Features
  • Unlike amber tinted tubes, these are completely opaque protecting even the most sensitive samples
  • RNase and DNase free
More About this Item
Black tubes provide a completely opaque barrier, protecting even the most light sensitive compounds. Ideal for use with fluorescent probes or ethidium bromide. Tubes are made of biocompatible polypropylene with a click-fit cap to prevent accidental opening. Tubes can be stored down to -100°C and can be autoclaved to 122°C.