This item is now discontinued. We recommend the following as suggested replacements: EW-01850-24
Argos Technologies Nova® Microcinerator, Loop and Needle Sterilizer; 220 VAC
Representative image only.
Argos Technologies Item # EW-04397-15
Reduce hand fatigue with easy-tilt burner head
  • Sterilize reusable loops, streakers, and needles without an open flame
  • Decontaminate platinum and nichrome wire inoculation instruments in 5 to 7 seconds
  • Sanitize safely—no gases, flames, or infectious spatter to worry about
  • Use as a slide warmer—optional adapter holds up to three slides while still allowing access to disinfect instruments
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Max Temperature (° F)1500
  • Max Temperature (° C)815
  • Average cycle time (minutes)0.1
  • Chamber constructionCeramic
  • Power (VAC)220
  • Power (Hz)50
  • DescriptionMicrocinerator, Loop and Needle Sterilizer; 220 VAC
  • Warranty1 Year
1 Year Warranty
Key Features
  • Sterilize reusable loops, streakers, and needles without an open flame
  • Decontaminate platinum and nichrome wire inoculation instruments in 5 to 7 seconds
  • Sanitize safely—no gases, flames, or infectious spatter to worry about
  • Use as a slide warmer—optional adapter holds up to three slides while still allowing access to disinfect instruments
  • 1 year warranty
More About this Item
The miniature incinerator warms up in 10 minutes and can be left on for long periods of time—perfect for a busy microbiology lab. Tilt the head to a position most comfortable for your application and use. Simply insert your inoculating instrument into the element half way, and hold it there for 5 to 7 seconds. No need to wait until the metal is glowing.
Base has small holes for holding your loops, streakers, and needles while not in use. Add the optional slide holder and warm up to three slides to help dry your cultures, or the optional loop rest to guide your instrument into the optimal sterilizing position and protect the handle from overheating. Slide warmer and loop rest may be used simultaneously.