Argos Technologies PolarSafe® 2D Storage Box, 81 Place for 5 mL Cryovials; 1/Ea
Representative image only.
Argos Technologies Item # EW-04395-85
Make sample processing even easier with automatic identification
  • Constructed of durable polycarbonate for cryogenic applications
  • Ideal for fast and accurate sample processing
Often Bought with...
SPECS & DESCRIPTION
Specifications & Description
  • Tubes Held81
  • Tube Size5mL
  • ColorBlue Grid
  • Description2D Storage Box, 81 Place for 5 mL Cryovials; 1/Ea
Key Features
  • Constructed of durable polycarbonate for cryogenic applications
  • Ideal for fast and accurate sample processing
More About this Item
The PolarSafe polycarbonate storage boxes work in conjuciton with the 2D tubes to provide automatic identification of both the box and stored vials. Each box features a 2D Datamatrix code on the bottom of the box as well as a laser engraved Code 128 barcode on the side. These boxes are available in "short" (for 2mL tubes) or "tall" designs (for 5mL tubes). White grid and blue grid options are available for quick and easy identification.