Argos Technologies SampleScan 96 Cryotube and Storage Box Barcode Scanner
Representative image only.
Argos Technologies Item # EW-04396-03
Instantly decodes 1D and 2D barcodes into any open application
  • Organize, track, and run QA/QC checks on tube samples
  • Works with all 2D and 1D marked microtubes, cryotubes, racks, plates, and storage boxes
  • Scans and decodes in less than one second into any open application
  • Powered by USB—installs in a few seconds for simple and flexible decoding of single tubes
  • Runs on Windows or Mac—no additional software is required for use
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithArgos Polarsafe 2D cryovials and stroage boxes
  • DescriptionCryotube and Storage Box Barcode Scanner
Key Features
  • Organize, track, and run QA/QC checks on tube samples
  • Works with all 2D and 1D marked microtubes, cryotubes, racks, plates, and storage boxes
  • Scans and decodes in less than one second into any open application
  • Powered by USB—installs in a few seconds for simple and flexible decoding of single tubes
  • Runs on Windows or Mac—no additional software is required for use
More About this Item
The Argos Technologies® cryogenic barcode scanner quickly scans 2D data matrix barcodes and 1D barcodes such as code 128, code 39, and more. Scan and decode in less than one second with no additional software required, simply output the scanned barcode directly into any open application accepting keyboard input. Simply hover a tube over the scanning window, or place it in the single tube holder, and the scanner sounds a beep and flashes an LED light to indicate that the scan was successful.
The compact footprint makes the device suitable even for crowded spaces, and enables efficient laboratory layout. The rugged metal chassis and glass scanning port allow for use in a range of lab environments. If moving tubes is not possible, the scanner can turn on its side to decode the barcode extending the utility of the device.
This scanner is compatible with all 2D and 1D marked labware from major manufacturers like Argos Technologies® PolarSafe®, Cole-Parmer® Essentials, Corning®, Nalgene™, Nunc, WHEATON®, and more! Help prepare or manage your laboratory's sample database.
SampleScan 96 can read the bar code on each tube in a 12-, 24-, 48-, 81-, 96-, 100-, or 384-place storage box. Reader features missing tube detection and a separate port to scan individual tubes instead of an entire rack at a time. Readings are completed in about 20 seconds per rack. Scanned data will output in a .csv file or to an open application for easy LIMS integration.