Argos Technologies Cell and Tissue Culture Plates, Nonsterile, Polystyrene, Untreated, 96-Well; 100/CS
Representative image only.
Argos Technologies Item # EW-24409-33
Optically clear plates allow for easy visualization of cell growth
  • Features untreated surface ideal for suspensions — does not facilitate cell attachment
  • Nonsterile
  • DNase/RNase free, nonpyrogenic, and noncytotoxic
$105.00USD / Case of 100
SPECS & DESCRIPTION
Specifications & Description
  • Number of Wells96
  • Min Working Well Volume (mL)0.075
  • Max Working Well Volume (mL)0.2
  • Well ShapeFlat
  • Culture Area (cm²)0.33
  • Surface TreatmentNo
  • Lid (Included)No
  • SterileNo
  • DescriptionCell and Tissue Culture Plates, Nonsterile, Polystyrene, Untreated, 96-Well; 100/CS
Key Features
  • Features untreated surface ideal for suspensions — does not facilitate cell attachment
  • Nonsterile
  • DNase/RNase free, nonpyrogenic, and noncytotoxic
More About this Item
Use these well plates to serve your various cell and tissue culture needs. Transparent polystyrene construction allows for easy visualization of cell growth and facilitates observation under a microscope. An alphanumeric notation on each well permits simple identification. These culture plates come without a lid, but can easily be covered with Cole-Parmer® Adhesive Microplate Sealing Film to help reduce evaporation and maximize results.
Manufactured under ISO 13485 and ISO 9001 quality management systems in an ISO Class 8 (formerly Class 100,000) clean room environment. These cell and tissue culture well plates are packaged 10 per bag. They are DNase/RNase free, non-pyrogenic, and non-cytotoxic; conform to USP Class VI; and are quality traceable with lot number. Compatible with most multiwell plate instruments and equipment.