This item is now discontinued. Please browse similar items
Argos Technologies Transfer Pipette, 250 µL, Exact Volume; 500/Bag
Representative image only.
Argos Technologies Item # EW-06226-07
No more breakage or cross-contamination
  • Translucent shatterproof LDPE is nontoxic and biologically inert
  • Nonbinding for cells and proteins to prevent sample loss
  • Heat sealable and liquid nitrogen compatible
  • Exact volume pipette is ideal for pecision applications and has a length of 102 mm
  • Designed with a 0.25 µL bulb draw and 23±1 drops per mL
SPECS & DESCRIPTION
Specifications & Description
  • SterileNo
  • AutoclavableNo
  • Max Volume (mL)0.25
  • Drops Per mL23
  • Overall Length (in)4.0625
  • Overall Length (mm)102
  • Tip StyleExact Volume
  • Body MaterialLow Density Polyethylene (LDPE)
  • Package Of500/Bag
  • DescriptionTransfer Pipette, 250 µL, Exact Volume; 500/Bag
Key Features
  • Translucent shatterproof LDPE is nontoxic and biologically inert
  • Nonbinding for cells and proteins to prevent sample loss
  • Heat sealable and liquid nitrogen compatible
  • Exact volume pipette is ideal for pecision applications and has a length of 102 mm
  • Designed with a 0.25 µL bulb draw and 23±1 drops per mL
More About this Item
Low-density polyethylene (LDPE) makes these disposable pipettes with built-in bulbs the perfect substitute for breakable glass pipettes, eliminating cross-contamination and cumbersome rubber bulb changes. Plastic reduces the loss of cells and proteins that often occurs with binding that takes place with glass alternatives. Precision manufacturing ensures uniform repeatable drops. Heat seal pipettes to provide liquid-tight transport and storage. These pipettes are suitable for use with a temperature range of -50 to 90°C and can be gas sterilized.