ResinTech Replacement Cartridge Kit for Hydra "Big Blue" High Capacity DI System
Representative image only.
ResinTech Mfr # HYK-122-B-HC – Item # EW-99285-18
Designed to keep your deionized water system running smoothly
  • Easily install and remove cartridges with quarter twist release canister
  • Made in the USA!
  • Compatible with Hydra DI deionized water system model 99285-14
  • Flow rates of up to 4.7 L/min.
  • 1 year warranty
PRODUCT OPTIONS
$504.12USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (grains)5710 as CaCO3
  • Max Inlet Pressure (PSI)100
  • Max Inlet Pressure (bar)6.9
  • Max Flow Rate (LPM)4.7
  • Diameter (in)4.5
  • DescriptionHigh Capacity Cartridge Replacement Kit for model 99285-14
  • Warranty1 year
1 Year Warranty
Key Features
  • Easily install and remove cartridges with quarter twist release canister
  • Made in the USA!
  • Compatible with Hydra DI deionized water system model 99285-14
  • Flow rates of up to 4.7 L/min.
  • 1 year warranty
More About this Item
Replace your cartridges regularly for best quality and performance. Water purification systems sold separately.
The high capacity cartridge replacement kit includes one 4.5" x 10" carbon GAC and two 4.5" x 20" high capacity cartridges.