Aries Filterworks Phoenix Biochem Remote Dispense System with Ultrafiltration module, 120V/60Hz

Representative image only.
Aries Filterworks Mfr # PHX-BC-R-1 – Item # EW-99285-34
Obtain Type I, 18.2 MΩ water—on demand!
  • Save money—best cost-of-ownership on the market
  • Save time—2.5 LPM flowrate and quick change of consumables
  • Built-in leak detection protects your investment
  • Full-color touch-screen display with intuitive interface

PRODUCT OPTIONS

$9,237.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • ASTM Water GradeType I
  • Max Flow Rate (L/min)2.5
  • Inlet Port3/8" OD tube
  • With UV LampNo
  • With UltrafilterYes
  • Resistivity Threshold18.2 MΩ-cm
  • Conductivity Threshold0.056 µS/cm
  • TOC Threshold5-10 ppb
  • Feed Water Requirements (Source)RO/DI/Distilled
  • Feed Water Requirements (Conductivity)<20 µS/cm
  • Feed Water Requirements (TOC)<50 ppb
  • Type I Water (Bacteria)<1 CFU/mL
  • Feed Water Requirements (pH range)5 to 8
  • Type I Water (Endotoxins)<0.005 EU/mL
  • Feed Water Requirements (pressure)20 psi (1.3 bar) min, 60 psi (4.1 bar) max
  • Feed Water Requirements (Temperature)100°F (37°C) max
  • Power (VAC)120
  • DescriptionBiochem Remote Dispense System with Ultrafiltration module, 120V/60Hz

Key Features

  • Save money—best cost-of-ownership on the market
  • Save time—2.5 LPM flowrate and quick change of consumables
  • Built-in leak detection protects your investment
  • Full-color touch-screen display with intuitive interface

More About this Item

The Phoenix systems use a staged purification process that includes high-purity ion exchange resins to remove dissolved minerals, and internal recirculation to further maintain water purity. Multiple sensors continuously monitor the system and final water quality to ensure trouble-free operation.

As water exits the system, a final filter removes particulates and bacteria to reach Type I water specifications. Additional options are available for special applications to include 185/254 nm UV, and or ultrafiltration (UF).

The Phoenix system is pre-configured into four application specific models with benchtop or remote dispesing options:
Phoenix Academy (models 99285-31 and -32): basic chemistry, academic, IC, and buffers
Phoenix Citation (models 99285-35 and -36 with UV): HPLC, GC-MS, and trace organics
Phoenix Biochem (models 99285-33 and -34 with UF): life science, cell culture, and microbiology
Phoenix Genome (models 99285-37 and -38 with UV and UF): DNA Sequencing, PCR, and electrophoresis

A complete package requires a system, conditioning cartridge, polishing cartridge, and final filter.
1. Choose base system
2. Choose conditioning cartridge 99285-45 for RO water feed, or 99285-46 for SDI/tap water feed
3. Choose polishing cartridge 99285-43 for standard results, or 99285-44 for additional organics removal
4. Choose final filter 99285-27 for up to 0.2µm pariculate removal, or 99285-47 to also include endotoxin removal

Options (must be ordered with unit, factory installation required):
Item 99285-39 - adds in flow totalizer function, viewable on display
Item 99285-40 - adds in a direct feed port
Item 99285-41 - adds in a recirculating dispensing gun (for units without a remote dispense function)
Item 99285-42 - adds in a wall mounting bracket to clear up bench space

Ideal for reagent grade solutions, life science, cell culture, microbiology, general water chemistry.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat