Aries Filterworks VP-17-3002 Filter Cartridge, Cation Hydrogen Form, Color Changing Resin, Standard Connection
Representative image only.
Aries Filterworks Mfr # VP-17-3002 – Item # EW-99284-75
Reduce operating costs while maintaining peak performance
  • High-capacity cartridges require fewer changes
  • Spin-welded construction with high-purity plastics ensures long service life
  • Available in standard connection and hose nipple configurations
  • Cation (hydrogen form), color changing Standard connection
PRODUCT OPTIONS
$96.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MediaCation Hydrogen Form, Color Changing
  • Capacity2975 grains
  • Capacity (grains)2975
  • ConnectionStandard 1" Top x 2" Bottom
  • Compatible withARIES, Unix, Barnstead, Vaponics
  • Max Inlet Pressure (PSI)30
  • Max Flow Rate (LPM)3.8
  • Max Flow Rate (GPH)60
  • Diameter (in)3.38
  • Length (in)17
  • DescriptionFilter Cartridge, Cation Hydrogen Form, Color Changing Resin, Standard Connection
Key Features
  • High-capacity cartridges require fewer changes
  • Spin-welded construction with high-purity plastics ensures long service life
  • Available in standard connection and hose nipple configurations
  • Cation (hydrogen form), color changing Standard connection
More About this Item
Use these filtration cartridges in your ARIES, UNIX, Barnstead® PCS® and Vaponics® water purification systems or design a system using wall-mounting brackets and extension tubing (order separately). Permanent joint welds create a hermetic seal and improve longevity in any installation. Nonpigmented ABS plastic housing is durable and produces high-purity laboratory-grade water. Choose between the standard ARIES connection type and the versatile 3/8" hose nipple, both of which are available in a variety of filter media.