This item is now discontinued. Please browse similar items

Ashcroft A230PSI Heavy Industrial Pressure Transmitter, 30 psig, 0.25%

Representative image only.
Ashcroft Mfr # A2-Y-A-M02-42-C1-30#-G – Item # EW-68455-41

SPECS & DESCRIPTION

Specifications & Description

  • Accuracy±0.25%
  • Min Pressure (PSI)0
  • Max Pressure (PSI)30
  • Process Connection1/4" NPT(M)
  • Electrical Connections1/2" NPT(M) conduit w/ pigtail
  • Output4 to 20 mA
  • DescriptionHeavy Industrial Pressure Transmitter, 30 psig, 0.25%

More About this Item

FEATURES

  • Choice of 1.0, 0.5 or 0.25% accuracy (% span)
  • Pressure ranges from 5 psi through 10,000 psi
  • CE mark is standard
  • 17-4 pH and 316L SS sensors available
  • 304 SS case in standard, welded or explosion proof construction
  • Designed for heavy industrial and hazardous location applications
  • Durable
  • Provides considerable specification flexibility in terms of performance, construction and optional features
  • Optional absolute pressure ranges available
  • Available externally adjustable zero and span access
  • FM, UL, CSA and ATEX listings available

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat