Ashcroft B Series Explosion Proof Pressure Switch, 100" WC, 1/4" NPT(F); Two Set Points
Representative image only.
Ashcroft Mfr # B724B100IW – Item # EW-68360-51
Switches are designed with a fixed set point and deadband
PRODUCT OPTIONS
$479.50USD / Each
$65.00 - $128.00USD / Each
for an additional
$65.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Max Pressure (in WC)100
  • CaseEpoxy-coated aluminum
  • Process Connection1/4" NPT(F)
  • Electrical Connections3/4" NPT(F) conduit
  • Enclosure RatingNEMA 7, 9, IP66
  • Media CompatibilityWater-based liquids compatible with Buna-N
  • DescriptionExplosion Proof Pressure Switch, 100" WC, 1/4" NPT(F); Two Set Points
More About this Item
Watertight general purpose pressure switches are ideal for industrial and OEM application.
Explosion proof pressure switches are designed for hazardous process and industrial applications with
ATEX and IECEx approval for hazardous locations.